The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297661.1 | NP_001284590.1 | 3 - 4 | 507 | 107 |
GenBank mRNA | BC050616.1 | - | 3 - 4 | 507 | 107 |