Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIP12
Entrez Gene ID
9320
Gene Name
thyroid hormone receptor interactor 12
Gene Aliases
TRIP-12, ULF
Chromosome Location
Chr.2: 229763837 - 229923234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284214.1 NP_001271143.1 19 - 20 2974 78
NM_001284215.1 NP_001271144.1 17 - 18 2799 78
NM_001284216.1 NP_001271145.1 16 - 17 2020 78
NM_004238.2 NP_004229.1 18 - 19 2830 78
XM_005246954.4 XP_005247011.1 19 - 20 3082 78
XM_005246955.4 XP_005247012.1 19 - 20 3081 78
XM_005246956.4 XP_005247013.1 19 - 20 3058 78
XM_005246957.4 XP_005247014.1 19 - 20 3048 78
XM_005246958.4 XP_005247015.1 19 - 20 3001 78
XM_005246960.4 XP_005247017.1 20 - 21 2995 78
XM_005246961.3 XP_005247018.1 18 - 19 2932 78
XM_005246962.4 XP_005247019.1 18 - 19 2929 78
XM_006712852.3 XP_006712915.1 18 - 19 2885 78
XM_011512180.2 XP_011510482.1 19 - 20 3035 78
XM_011512181.2 XP_011510483.1 19 - 20 3033 78
XM_011512183.2 XP_011510485.1 18 - 19 2889 78
XM_017005272.1 XP_016860761.1 20 - 21 3141 78
XM_017005273.1 XP_016860762.1 19 - 20 3008 78
XM_017005274.1 XP_016860763.1 19 - 20 2956 78
XM_017005275.1 XP_016860764.1 19 - 20 3011 78
XM_017005276.1 XP_016860765.1 19 - 20 3016 78
XM_017005277.1 XP_016860766.1 20 - 21 3156 78
XM_017005278.1 XP_016860767.1 19 - 20 2971 78
XM_017005279.1 XP_016860768.1 19 - 20 3043 78
XM_017005280.1 XP_016860769.1 19 - 20 3032 78
XM_017005281.1 XP_016860770.1 19 - 20 2951 78
XM_017005282.1 XP_016860771.1 20 - 21 2994 78
XM_017005283.1 XP_016860772.1 19 - 20 2998 78
XM_017005284.1 XP_016860773.1 20 - 21 2992 78
XM_017005285.1 XP_016860774.1 18 - 19 2919 78
XM_017005286.1 XP_016860775.1 18 - 19 2951 78
XM_017005287.1 XP_016860776.1 18 - 19 2932 78
XM_017005288.1 XP_016860777.1 18 - 19 2894 78
XM_017005289.1 XP_016860778.1 20 - 21 2914 78
XM_017005290.1 XP_016860779.1 20 - 21 2911 78
XM_017005291.1 XP_016860780.1 19 - 20 2845 78
XM_017005292.1 XP_016860781.1 18 - 19 2848 78
XM_017005293.1 XP_016860782.1 18 - 19 2848 78
GenBank mRNA BC113891.1 - 16 - 17 1914 78
BC114556.1 - 19 - 20 2868 78
D28476.1 - 18 - 19 2760 78
EU489742.1 - 17 - 18 2753 78

View Full Product Details