Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIP12
Entrez Gene ID
9320
Gene Name
thyroid hormone receptor interactor 12
Gene Aliases
TRIP-12, ULF
Chromosome Location
Chr.2: 229763837 - 229923234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284214.1 NP_001271143.1 27 - 28 4240 77
NM_001284215.1 NP_001271144.1 25 - 26 4065 77
NM_001284216.1 NP_001271145.1 24 - 25 3286 77
NM_004238.2 NP_004229.1 26 - 27 4096 77
XM_005246954.4 XP_005247011.1 27 - 28 4348 77
XM_005246955.4 XP_005247012.1 27 - 28 4347 77
XM_005246956.4 XP_005247013.1 27 - 28 4324 77
XM_005246957.4 XP_005247014.1 27 - 28 4299 77
XM_005246958.4 XP_005247015.1 27 - 28 4267 77
XM_005246960.4 XP_005247017.1 28 - 29 4261 77
XM_005246961.3 XP_005247018.1 26 - 27 4198 77
XM_005246962.4 XP_005247019.1 26 - 27 4195 77
XM_006712852.3 XP_006712915.1 26 - 27 4151 77
XM_011512180.2 XP_011510482.1 27 - 28 4301 77
XM_011512181.2 XP_011510483.1 27 - 28 4299 77
XM_011512183.2 XP_011510485.1 26 - 27 4155 77
XM_017005272.1 XP_016860761.1 28 - 29 4407 77
XM_017005273.1 XP_016860762.1 27 - 28 4274 77
XM_017005274.1 XP_016860763.1 27 - 28 4222 77
XM_017005275.1 XP_016860764.1 27 - 28 4277 77
XM_017005276.1 XP_016860765.1 27 - 28 4282 77
XM_017005277.1 XP_016860766.1 28 - 29 4422 77
XM_017005278.1 XP_016860767.1 27 - 28 4237 77
XM_017005279.1 XP_016860768.1 27 - 28 4294 77
XM_017005280.1 XP_016860769.1 27 - 28 4283 77
XM_017005281.1 XP_016860770.1 27 - 28 4217 77
XM_017005282.1 XP_016860771.1 28 - 29 4260 77
XM_017005283.1 XP_016860772.1 27 - 28 4249 77
XM_017005284.1 XP_016860773.1 28 - 29 4243 77
XM_017005285.1 XP_016860774.1 26 - 27 4185 77
XM_017005286.1 XP_016860775.1 26 - 27 4217 77
XM_017005287.1 XP_016860776.1 26 - 27 4183 77
XM_017005288.1 XP_016860777.1 26 - 27 4145 77
XM_017005289.1 XP_016860778.1 28 - 29 4180 77
XM_017005290.1 XP_016860779.1 28 - 29 4177 77
XM_017005291.1 XP_016860780.1 27 - 28 4111 77
XM_017005292.1 XP_016860781.1 26 - 27 4099 77
XM_017005293.1 XP_016860782.1 26 - 27 4114 77
GenBank mRNA BC113891.1 - 24 - 25 3180 77
BC114556.1 - 27 - 28 4134 77
D28476.1 - 26 - 27 4026 77
EU489742.1 - 25 - 26 4019 77

View Full Product Details