Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIP12
Entrez Gene ID
9320
Gene Name
thyroid hormone receptor interactor 12
Gene Aliases
TRIP-12, ULF
Chromosome Location
Chr.2: 229763837 - 229923234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284214.1 NP_001271143.1 32 - 33 4937 96
NM_001284215.1 NP_001271144.1 30 - 31 4762 96
NM_001284216.1 NP_001271145.1 29 - 30 3983 96
NM_004238.2 NP_004229.1 31 - 32 4793 96
XM_005246954.4 XP_005247011.1 32 - 33 5045 96
XM_005246955.4 XP_005247012.1 32 - 33 5044 96
XM_005246956.4 XP_005247013.1 32 - 33 5021 96
XM_005246957.4 XP_005247014.1 32 - 33 4996 96
XM_005246958.4 XP_005247015.1 32 - 33 4964 96
XM_005246960.4 XP_005247017.1 33 - 34 4958 96
XM_005246961.3 XP_005247018.1 31 - 32 4895 96
XM_005246962.4 XP_005247019.1 31 - 32 4892 96
XM_006712852.3 XP_006712915.1 31 - 32 4848 96
XM_011512180.2 XP_011510482.1 32 - 33 4998 96
XM_011512181.2 XP_011510483.1 32 - 33 4996 96
XM_011512183.2 XP_011510485.1 31 - 32 4852 96
XM_017005272.1 XP_016860761.1 33 - 34 5104 96
XM_017005273.1 XP_016860762.1 32 - 33 4971 96
XM_017005274.1 XP_016860763.1 32 - 33 4919 96
XM_017005275.1 XP_016860764.1 32 - 33 4974 96
XM_017005276.1 XP_016860765.1 32 - 33 4979 96
XM_017005277.1 XP_016860766.1 33 - 34 5119 96
XM_017005278.1 XP_016860767.1 32 - 33 4934 96
XM_017005279.1 XP_016860768.1 32 - 33 4991 96
XM_017005280.1 XP_016860769.1 32 - 33 4980 96
XM_017005281.1 XP_016860770.1 32 - 33 4914 96
XM_017005282.1 XP_016860771.1 33 - 34 4957 96
XM_017005283.1 XP_016860772.1 32 - 33 4946 96
XM_017005284.1 XP_016860773.1 33 - 34 4940 96
XM_017005285.1 XP_016860774.1 31 - 32 4882 96
XM_017005286.1 XP_016860775.1 31 - 32 4914 96
XM_017005287.1 XP_016860776.1 31 - 32 4880 96
XM_017005288.1 XP_016860777.1 31 - 32 4842 96
XM_017005289.1 XP_016860778.1 33 - 34 4877 96
XM_017005290.1 XP_016860779.1 33 - 34 4874 96
XM_017005291.1 XP_016860780.1 32 - 33 4808 96
XM_017005292.1 XP_016860781.1 31 - 32 4796 96
XM_017005293.1 XP_016860782.1 31 - 32 4811 96
GenBank mRNA BC113891.1 - 29 - 30 3877 96
BC114556.1 - 32 - 33 4831 96
D28476.1 - 31 - 32 4723 96
EU489742.1 - 30 - 31 4716 96

View Full Product Details