The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001137604.2 | NP_001131076.1 | 2 - 3 | 902 | 96 |
NM_001282772.1 | NP_001269701.1 | 4 - 5 | 720 | 96 | |
NM_001282774.1 | NP_001269703.1 | 3 - 4 | 1070 | 96 | |
NM_001282777.1 | NP_001269706.1 | 4 - 5 | 1162 | 96 | |
NM_001282779.1 | NP_001269708.1 | 3 - 4 | 999 | 96 | |
NM_019014.5 | NP_061887.2 | 3 - 4 | 1070 | 96 | |
GenBank mRNA | AK001678.1 | - | 3 - 4 | 542 | 96 |
AK022533.1 | - | 2 - 3 | 352 | 96 | |
AK128044.1 | - | 3 - 4 | 1070 | 96 | |
AK294027.1 | - | 3 - 4 | 471 | 96 | |
AK302814.1 | - | 4 - 5 | 720 | 96 | |
BC110833.1 | - | 3 - 4 | 759 | 96 | |
BX647360.1 | - | 4 - 5 | 888 | 96 | |
BX647683.1 | - | 3 - 4 | 449 | 96 |