The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198810.1 | NP_001185739.1 | 3 - 4 | 446 | 70 |
NM_003627.5 | NP_003618.1 | 3 - 4 | 617 | 70 | |
XM_005274358.4 | XP_005274415.1 | 4 - 5 | 757 | 70 | |
XM_011545320.2 | XP_011543622.1 | 4 - 5 | 759 | 70 | |
XM_011545321.2 | XP_011543623.1 | 3 - 4 | 460 | 70 | |
XM_011545322.1 | XP_011543624.1 | 2 - 3 | 309 | 70 | |
XM_017018451.1 | XP_016873940.1 | 4 - 5 | 645 | 70 | |
XM_017018452.1 | XP_016873941.1 | 3 - 4 | 679 | 70 | |
XM_017018453.1 | XP_016873942.1 | 3 - 4 | 650 | 70 | |
GenBank mRNA | AB103033.1 | - | 3 - 4 | 584 | 70 |
AF045584.1 | - | 3 - 4 | 404 | 70 | |
AK027732.1 | - | 1 - 2 | 182 | 70 | |
BC001639.2 | - | 3 - 4 | 446 | 70 |