Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL24
Entrez Gene ID
54800
Gene Name
kelch like family member 24
Gene Aliases
DRE1, KRIP6
Chromosome Location
Chr.3: 183635117 - 183684519 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017644.3 NP_060114.2 7 - 8 1900 98
XM_005247552.2 XP_005247609.1 8 - 9 1991 98
XM_005247553.1 XP_005247610.1 8 - 9 1945 98
XM_005247554.2 XP_005247611.1 7 - 8 1856 98
XM_005247555.1 XP_005247612.1 7 - 8 1928 98
XM_005247556.1 XP_005247613.1 6 - 7 1850 98
XM_011512938.2 XP_011511240.1 6 - 7 1792 98
XM_017006650.1 XP_016862139.1 8 - 9 2481 98
XM_017006651.1 XP_016862140.1 9 - 10 2066 98
XM_017006652.1 XP_016862141.1 9 - 10 2020 98
XM_017006653.1 XP_016862142.1 8 - 9 1942 98
XM_017006654.1 XP_016862143.1 8 - 9 1932 98
XM_017006655.1 XP_016862144.1 8 - 9 2003 98
XM_017006656.1 XP_016862145.1 7 - 8 1925 98
XM_017006657.1 XP_016862146.1 8 - 9 1957 98
XM_017006658.1 XP_016862147.1 7 - 8 1866 98
XM_017006659.1 XP_016862148.1 8 - 9 3692 98
XM_017006660.1 XP_016862149.1 7 - 8 1882 98
XM_017006661.1 XP_016862150.1 10 - 11 2183 98
XM_017006662.1 XP_016862151.1 8 - 9 2039 98
XM_017006663.1 XP_016862152.1 8 - 9 2028 98
GenBank mRNA AK026326.1 - 5 - 6 1309 98
AY422472.1 - 7 - 8 1900 98
BX648466.1 - 8 - 9 1992 98
BX648812.1 - 8 - 9 1992 98
DQ925701.1 - 5 - 6 1605 98

View Full Product Details