The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288985.1 | NP_001275914.1 | 21 - 22 | 3103 | 75 |
NM_001288986.1 | NP_001275915.1 | 20 - 21 | 2942 | 75 | |
NM_007168.3 | NP_009099.1 | 19 - 20 | 2822 | 75 | |
XM_005256938.2 | XP_005256995.1 | 20 - 21 | 2970 | 75 | |
XM_005256940.4 | XP_005256997.1 | 19 - 20 | 2875 | 75 | |
XM_011524191.2 | XP_011522493.1 | 20 - 21 | 2933 | 75 | |
XM_011524192.2 | XP_011522494.1 | 20 - 21 | 2933 | 75 | |
GenBank mRNA | AB020629.1 | - | 19 - 20 | 2781 | 75 |
AK295876.1 | - | 20 - 21 | 2915 | 75 | |
AK304286.1 | - | 19 - 20 | 2774 | 75 | |
BC037309.1 | - | 21 - 22 | 2870 | 75 | |
BC130280.1 | - | 21 - 22 | 2952 | 75 |