The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002758.3 | NP_002749.2 | 4 - 5 | 534 | 65 |
XM_005257515.2 | XP_005257572.1 | 4 - 5 | 598 | 65 | |
XM_005257516.2 | XP_005257573.1 | 4 - 5 | 471 | 65 | |
XM_006721975.3 | XP_006722038.1 | 4 - 5 | 499 | 65 | |
XM_011525025.2 | XP_011523327.1 | 4 - 5 | 885 | 65 | |
XM_011525026.2 | XP_011523328.1 | 4 - 5 | 346 | 65 | |
XM_011525027.2 | XP_011523329.1 | 4 - 5 | 261 | 65 | |
GenBank mRNA | AB451252.1 | - | 4 - 5 | 246 | 65 |
AB451376.1 | - | 4 - 5 | 246 | 65 | |
AK225719.1 | - | 4 - 5 | 528 | 65 | |
AK290747.1 | - | 4 - 5 | 546 | 65 | |
BC012009.1 | - | 4 - 5 | 444 | 65 | |
BX641121.1 | - | 3 - 4 | 595 | 65 | |
D87905.1 | - | 4 - 5 | 246 | 65 | |
U39064.1 | - | 4 - 5 | 482 | 65 | |
U39065.1 | - | 4 - 5 | 531 | 65 | |
U39656.1 | - | 4 - 5 | 586 | 65 | |
U39657.1 | - | 4 - 5 | 586 | 65 | |
U49732.1 | - | 4 - 5 | 491 | 65 | |
X96757.1 | - | 4 - 5 | 518 | 65 |