The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030783.1 | NP_110410.1 | 4 - 5 | 607 | 137 |
XM_011520392.2 | XP_011518694.1 | 5 - 6 | 604 | 137 | |
XM_011520393.2 | XP_011518695.1 | 3 - 4 | 322 | 137 | |
XM_017018368.1 | XP_016873857.1 | 6 - 7 | 2999 | 137 | |
XM_017018369.1 | XP_016873858.1 | 5 - 6 | 729 | 137 | |
XM_017018370.1 | XP_016873859.1 | 3 - 4 | 404 | 137 | |
XM_017018371.1 | XP_016873860.1 | 4 - 5 | 518 | 137 | |
GenBank mRNA | AL834357.1 | - | 4 - 5 | 521 | 137 |
BC001210.1 | - | 4 - 5 | 607 | 137 |