The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302623.1 | NP_001289552.1 | 3 - 4 | 426 | 117 |
NM_001322216.1 | NP_001309145.1 | 3 - 4 | 478 | 117 | |
NM_001322217.1 | NP_001309146.1 | 2 - 3 | 392 | 117 | |
NM_001322218.1 | NP_001309147.1 | 3 - 4 | 478 | 117 | |
NM_001322219.1 | NP_001309148.1 | 3 - 4 | 478 | 117 | |
NM_001322220.1 | NP_001309149.1 | 3 - 4 | 478 | 117 | |
NM_001322221.1 | NP_001309150.1 | 3 - 4 | 478 | 117 | |
NM_004941.2 | NP_004932.1 | 3 - 4 | 426 | 117 | |
NR_136225.1 | 3 - 4 | 478 | 117 | ||
NR_136226.1 | 3 - 4 | 478 | 117 | ||
NR_136227.1 | 3 - 4 | 478 | 117 | ||
NR_136228.1 | 3 - 4 | 478 | 117 | ||
GenBank mRNA | AK291664.1 | - | 3 - 4 | 359 | 117 |
AK294046.1 | - | 2 - 3 | 392 | 117 | |
AK303319.1 | - | 3 - 4 | 380 | 117 | |
BC044586.1 | - | 3 - 4 | 324 | 117 | |
BC047327.1 | - | 3 - 4 | 328 | 117 | |
D50487.1 | - | 3 - 4 | 378 | 117 | |
DC310785.1 | - | 2 - 3 | 392 | 117 |