Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK8
Entrez Gene ID
1024
Gene Name
cyclin dependent kinase 8
Gene Aliases
K35
Chromosome Location
Chr.13: 26254104 - 26405238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260.2 NP_001251.1 10 - 11 1570 78
NM_001318368.1 NP_001305297.1 10 - 11 1570 78
XM_005266216.4 XP_005266273.1 10 - 11 928 78
XM_011534864.1 XP_011533166.1 9 - 10 1491 78
XM_011534865.2 XP_011533167.1 6 - 7 1615 78
XM_017020317.1 XP_016875806.1 9 - 10 926 78
XM_017020318.1 XP_016875807.1 6 - 7 660 78
GenBank mRNA AK296084.1 - 6 - 7 560 78
AK296875.1 - 9 - 10 913 78
AY702976.1 - 9 - 10 974 78
BC069634.1 - 10 - 11 1055 78
BC094784.1 - 9 - 10 1512 78
BC104492.1 - 10 - 11 1058 78
BC105950.1 - 10 - 11 1058 78
BC107601.1 - 10 - 11 1057 78
L23208.1 - 2 - 3 168 78
X85753.1 - 10 - 11 1058 78

Gene Symbol
ADRA1B
Entrez Gene ID
147
Gene Name
adrenoceptor alpha 1B
Gene Aliases
ADRA1, ALPHA1BAR
Chromosome Location
Chr.5: 159916733 - 159989205 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK309989.1 - - 1291 78

View Full Product Details