The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172713.1 | NP_001166184.1 | 14 - 15 | 2122 | 125 |
NM_002078.4 | NP_002069.2 | 13 - 14 | 2056 | 125 | |
XM_005265069.3 | XP_005265126.1 | 15 - 16 | 2141 | 125 | |
XM_005265070.3 | XP_005265127.1 | 15 - 16 | 2141 | 125 | |
XM_005265071.3 | XP_005265128.1 | 15 - 16 | 2141 | 125 | |
XM_005265072.3 | XP_005265129.1 | 14 - 15 | 2093 | 125 | |
XM_005265073.1 | XP_005265130.1 | 14 - 15 | 2042 | 125 | |
XM_005265074.1 | XP_005265131.1 | 14 - 15 | 2042 | 125 | |
XM_005265075.1 | XP_005265132.1 | 12 - 13 | 1930 | 125 | |
XM_006713110.2 | XP_006713173.1 | 13 - 14 | 2036 | 125 | |
XM_011533598.2 | XP_011531900.1 | 9 - 10 | 1515 | 125 | |
XM_017006184.1 | XP_016861673.1 | 15 - 16 | 2141 | 125 | |
XM_017006185.1 | XP_016861674.1 | 15 - 16 | 2631 | 125 | |
XM_017006186.1 | XP_016861675.1 | 15 - 16 | 1810 | 125 | |
XM_017006187.1 | XP_016861676.1 | 13 - 14 | 1994 | 125 | |
XM_017006188.1 | XP_016861677.1 | 11 - 12 | 1873 | 125 | |
GenBank mRNA | AB209693.1 | - | 14 - 15 | 2037 | 125 |
AF273047.1 | - | 7 - 8 | 914 | 125 | |
AK290550.1 | - | 13 - 14 | 1978 | 125 | |
AK307429.1 | - | 4 - 5 | 659 | 125 | |
U31906.1 | - | 11 - 12 | 1310 | 125 | |
U41740.1 | - | 13 - 14 | 1981 | 125 | |
X76942.1 | - | 1 - 2 | 129 | 125 | |
X82834.1 | - | 13 - 14 | 1903 | 125 |