The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080283.3 | NP_525022.2 | 21 - 22 | 3047 | 117 |
XM_017024011.1 | XP_016879500.1 | 21 - 22 | 3012 | 117 | |
XM_017024012.1 | XP_016879501.1 | 21 - 22 | 3116 | 117 | |
XM_017024013.1 | XP_016879502.1 | 21 - 22 | 2936 | 117 | |
XM_017024014.1 | XP_016879503.1 | 21 - 22 | 3536 | 117 | |
XM_017024015.1 | XP_016879504.1 | 18 - 19 | 2559 | 117 | |
GenBank mRNA | AF423307.1 | - | 20 - 21 | 2904 | 117 |
AY028899.1 | - | 21 - 22 | 3047 | 117 | |
CR627382.1 | - | 20 - 21 | 2853 | 117 |