Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFEC
Entrez Gene ID
22797
Gene Name
transcription factor EC
Gene Aliases
TCFEC, TFE-C, TFEC-L, TFECL, bHLHe34, hTFEC-L
Chromosome Location
Chr.7: 115935148 - 116159891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018058.2 NP_001018068.1 5 - 6 685 92
NM_001244583.1 NP_001231512.1 4 - 5 381 92
NM_012252.3 NP_036384.1 6 - 7 772 92
XM_011515963.1 XP_011514265.1 8 - 9 959 92
XM_011515964.2 XP_011514266.1 8 - 9 974 92
XM_011515965.2 XP_011514267.1 8 - 9 914 92
XM_011515966.2 XP_011514268.1 8 - 9 1023 92
XM_011515969.1 XP_011514271.1 7 - 8 902 92
XM_011515970.1 XP_011514272.1 7 - 8 872 92
XM_011515972.2 XP_011514274.1 5 - 6 488 92
XM_017011874.1 XP_016867363.1 7 - 8 1075 92
XM_017011875.1 XP_016867364.1 7 - 8 910 92
XM_017011876.1 XP_016867365.1 4 - 5 347 92
XM_017011877.1 XP_016867366.1 6 - 7 736 92
XM_017011878.1 XP_016867367.1 4 - 5 274 92
GenBank mRNA AK301479.1 - 8 - 9 964 92
AK313546.1 - 6 - 7 724 92
BC029891.1 - 5 - 6 616 92
CR933605.1 - 5 - 6 685 92
D43945.1 - 6 - 7 701 92

View Full Product Details