The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251989.1 | NP_001238918.1 | 3 - 4 | 338 | 138 |
NM_001251990.1 | NP_001238919.1 | 3 - 4 | 262 | 138 | |
NM_001251991.1 | NP_001238920.1 | 3 - 4 | 246 | 138 | |
NM_015895.4 | NP_056979.1 | 3 - 4 | 470 | 138 | |
XM_005249159.1 | XP_005249216.1 | 3 - 4 | 509 | 138 | |
XM_011514651.2 | XP_011512953.1 | 2 - 3 | 2564 | 138 | |
GenBank mRNA | AF067855.1 | - | 2 - 3 | 156 | 138 |
AK021685.1 | - | 3 - 4 | 338 | 138 | |
AK222992.1 | - | 2 - 3 | 129 | 138 | |
AK315743.1 | - | 3 - 4 | 451 | 138 | |
BC005185.1 | - | 3 - 4 | 393 | 138 | |
BC005389.1 | - | 3 - 4 | 445 | 138 | |
BG777134.1 | - | 3 - 4 | 218 | 138 | |
BM699599.1 | - | 3 - 4 | 244 | 138 | |
CB992796.1 | - | 3 - 4 | 357 | 138 |