Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX14
Entrez Gene ID
5195
Gene Name
peroxisomal biogenesis factor 14
Gene Aliases
NAPP2, PBD13A, Pex14p, dJ734G22.2
Chromosome Location
Chr.1: 10474946 - 10630758 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004565.2 NP_004556.1 6 - 7 511 102
XM_005263470.4 XP_005263527.1 7 - 8 3663 102
XM_011541577.2 XP_011539879.1 6 - 7 605 102
XM_011541578.2 XP_011539880.1 6 - 7 862 102
XM_011541579.2 XP_011539881.1 5 - 6 942 102
XM_011541580.1 XP_011539882.1 5 - 6 382 102
GenBank mRNA AB017546.1 - 6 - 7 495 102
AF045186.1 - 6 - 7 495 102
AK002194.1 - 6 - 7 503 102
AK293684.1 - 5 - 6 386 102
AK298136.1 - 5 - 6 421 102
AK307416.1 - 6 - 7 465 102
AK307462.1 - 5 - 6 511 102
AK313046.1 - 6 - 7 495 102
BC006327.2 - 6 - 7 502 102
CR542083.1 - 6 - 7 490 102

View Full Product Details