Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBPL6
Entrez Gene ID
114880
Gene Name
oxysterol binding protein like 6
Gene Aliases
ORP6
Chromosome Location
Chr.2: 178194479 - 178399433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201480.1 NP_001188409.1 4 - 5 737 104
NM_001201481.1 NP_001188410.1 4 - 5 737 104
NM_001201482.1 NP_001188411.1 4 - 5 737 104
NM_032523.3 NP_115912.1 4 - 5 737 104
NM_145739.2 NP_665682.1 2 - 3 250 104
XM_017003265.1 XP_016858754.1 5 - 6 809 104
XM_017003266.1 XP_016858755.1 5 - 6 809 104
XM_017003267.1 XP_016858756.1 3 - 4 504 104
XM_017003268.1 XP_016858757.1 4 - 5 733 104
XM_017003269.1 XP_016858758.1 3 - 4 504 104
XM_017003270.1 XP_016858759.1 5 - 6 891 104
XM_017003271.1 XP_016858760.1 3 - 4 504 104
XM_017003274.1 XP_016858763.1 4 - 5 733 104
XM_017003275.1 XP_016858764.1 5 - 6 827 104
GenBank mRNA AB208898.1 - 4 - 5 737 104
AF323728.1 - 4 - 5 571 104
AF392448.1 - 4 - 5 571 104
AF462443.1 - 3 - 4 547 104
AK093902.1 - 2 - 3 252 104
AK300389.1 - 4 - 5 695 104
BC052259.1 - 4 - 5 718 104

View Full Product Details