The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307944.1 | NP_001294873.1 | 6 - 7 | 1072 | 73 |
GenBank mRNA | AK056354.1 | - | 6 - 7 | 1041 | 73 |