Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABTB1
Entrez Gene ID
80325
Gene Name
ankyrin repeat and BTB domain containing 1
Gene Aliases
BPOZ, BTB3, BTBD21, EF1ABP, PP2259
Chromosome Location
Chr.3: 127672928 - 127680926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032548.3 NP_115937.1 11 - 12 1323 72
NM_172027.2 NP_742024.1 11 - 12 1312 72
NR_033429.1 11 - 12 1277 72
XM_006713769.3 XP_006713832.1 11 - 12 1346 72
XM_011513213.1 XP_011511515.1 11 - 12 1246 72
XM_017007285.1 XP_016862774.1 10 - 11 1175 72
XM_017007286.1 XP_016862775.1 10 - 11 1157 72
XM_017007287.1 XP_016862776.1 11 - 12 1283 72
XM_017007288.1 XP_016862777.1 10 - 11 1216 72
GenBank mRNA AB053324.1 - 11 - 12 1302 72
AB053325.1 - 11 - 12 1291 72
AB053326.1 - 11 - 12 1256 72
AF297986.1 - 11 - 12 1269 72
AF447886.1 - 10 - 11 1416 72
AK090509.1 - 1 - 2 1684 72
AK098738.1 - 4 - 5 443 72
AK130594.1 - 7 - 8 802 72
BC011858.2 - 11 - 12 1310 72

View Full Product Details