The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032548.3 | NP_115937.1 | 8 - 9 | 862 | 83 |
NM_172027.2 | NP_742024.1 | 8 - 9 | 851 | 83 | |
NR_033429.1 | 8 - 9 | 816 | 83 | ||
XM_006713769.3 | XP_006713832.1 | 8 - 9 | 885 | 83 | |
XM_011513213.1 | XP_011511515.1 | 8 - 9 | 785 | 83 | |
XM_017007285.1 | XP_016862774.1 | 7 - 8 | 714 | 83 | |
XM_017007286.1 | XP_016862775.1 | 7 - 8 | 696 | 83 | |
XM_017007287.1 | XP_016862776.1 | 8 - 9 | 822 | 83 | |
XM_017007288.1 | XP_016862777.1 | 7 - 8 | 755 | 83 | |
GenBank mRNA | AB053324.1 | - | 8 - 9 | 841 | 83 |
AB053325.1 | - | 8 - 9 | 830 | 83 | |
AB053326.1 | - | 8 - 9 | 795 | 83 | |
AF297986.1 | - | 8 - 9 | 808 | 83 | |
AF447886.1 | - | 7 - 8 | 955 | 83 | |
AK090422.1 | - | 2 - 3 | 1698 | 83 | |
AK315148.1 | - | 8 - 9 | 831 | 83 | |
BC011858.2 | - | 8 - 9 | 849 | 83 |