The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144952.1 | NP_001138424.1 | 32 - 33 | 4569 | 63 |
XM_011524914.2 | XP_011523216.1 | 32 - 33 | 4953 | 63 | |
XM_011524915.2 | XP_011523217.1 | 32 - 33 | 4955 | 63 | |
GenBank mRNA | AB040947.1 | - | 25 - 26 | 3600 | 63 |
AL137357.1 | - | 10 - 11 | 1477 | 63 | |
BC045763.1 | - | 14 - 15 | 2097 | 63 | |
BC066363.1 | - | 24 - 25 | 3469 | 63 |