The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007015.2 | NP_008946.1 | 6 - 7 | 905 | 73 |
XM_011534897.2 | XP_011533199.1 | 6 - 7 | 915 | 73 | |
XM_011534899.2 | XP_011533201.1 | 5 - 6 | 803 | 73 | |
XM_011534900.2 | XP_011533202.1 | 5 - 6 | 789 | 73 | |
GenBank mRNA | AB005999.1 | - | 1 - 2 | 152 | 73 |
AB006000.1 | - | 6 - 7 | 794 | 73 | |
CR541910.1 | - | 6 - 7 | 794 | 73 |