The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016001.2 | NP_057085.2 | 11 - 12 | 1553 | 141 |
GenBank mRNA | AF151806.1 | - | 11 - 12 | 1539 | 141 |
AK026355.1 | - | 12 - 13 | 1302 | 141 | |
AK314399.1 | - | 11 - 12 | 1532 | 141 | |
AY007138.1 | - | 9 - 10 | 1023 | 141 | |
BC025276.1 | - | 11 - 12 | 1529 | 141 | |
BM548967.1 | - | 3 - 4 | 141 |