Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXOSC3
Entrez Gene ID
51010
Gene Name
exosome component 3
Gene Aliases
CGI-102, PCH1B, RRP40, Rrp40p, bA3J10.7, hRrp-40, p10
Chromosome Location
Chr.9: 37779714 - 37785092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016042.3 NP_057126.2 2 - 3 523 150
GenBank mRNA AF151860.1 - 2 - 3 449 150
AF281132.1 - 2 - 3 499 150
AK290864.1 - 2 - 3 488 150
BC002437.2 - 2 - 3 487 150
BC008880.2 - 2 - 3 477 150

View Full Product Details