Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
-
Chromosome Location
Chr.17: 44141902 - 44162477 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171251.2 NP_001164722.1 4 - 5 1546 61
NM_001321311.1 NP_001308240.1 4 - 5 1543 61
NM_024032.4 NP_076937.2 4 - 5 1546 61
XM_011525187.1 XP_011523489.1 4 - 5 1393 61
XM_011525191.2 XP_011523493.1 4 - 5 1396 61
XM_011525192.1 XP_011523494.1 4 - 5 1396 61
XM_011525194.1 XP_011523496.1 4 - 5 1396 61
XM_011525195.1 XP_011523497.1 4 - 5 1396 61
XM_017025031.1 XP_016880520.1 4 - 5 1569 61
XM_017025032.1 XP_016880521.1 4 - 5 1568 61
GenBank mRNA AK291924.1 - 4 - 5 1540 61
AL834295.1 - 4 - 5 1490 61

View Full Product Details