The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161585.1 | NP_001155057.1 | 1 - 2 | 162 | 121 |
NM_020641.2 | NP_065692.2 | 1 - 2 | 162 | 121 | |
GenBank mRNA | AJ278482.1 | - | 1 - 2 | 162 | 121 |
BC014307.1 | - | 1 - 2 | 162 | 121 | |
BC137351.1 | - | 1 - 2 | 132 | 121 | |
BC137352.1 | - | 1 - 2 | 132 | 121 | |
BC144602.1 | - | 1 - 2 | 132 | 121 |