Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBN2
Entrez Gene ID
2201
Gene Name
fibrillin 2
Gene Aliases
CCA, DA9, EOMD
Chromosome Location
Chr.5: 128257908 - 128538358 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001999.3 NP_001990.2 22 - 23 3296 65
XM_017009228.1 XP_016864717.1 21 - 22 3459 65
GenBank mRNA AB209735.1 - 1 - 2 49 65
AK300440.1 - 21 - 22 2976 65
U03272.1 - 22 - 23 2854 65
X62009.1 - 6 - 7 601 65

View Full Product Details