The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001999.3 | NP_001990.2 | 30 - 31 | 4404 | 103 |
XM_017009228.1 | XP_016864717.1 | 29 - 30 | 4567 | 103 | |
GenBank mRNA | AB209735.1 | - | 9 - 10 | 1157 | 103 |
AK300440.1 | - | 29 - 30 | 4084 | 103 | |
U03272.1 | - | 30 - 31 | 3962 | 103 | |
X62009.1 | - | 14 - 15 | 1709 | 103 |