The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001999.3 | NP_001990.2 | 63 - 64 | 8630 | 109 |
XM_017009228.1 | XP_016864717.1 | 62 - 63 | 8793 | 109 | |
GenBank mRNA | AB209735.1 | - | 42 - 43 | 5383 | 109 |
CN300904.1 | - | 4 - 5 | 535 | 109 | |
U03272.1 | - | 63 - 64 | 8188 | 109 |