The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278731.1 | NP_001265660.1 | 9 - 10 | 1346 | 73 |
NM_001278732.1 | NP_001265661.1 | 8 - 9 | 1299 | 73 | |
NM_012208.3 | NP_036340.1 | 10 - 11 | 1421 | 73 | |
XM_011537619.1 | XP_011535921.1 | 11 - 12 | 1434 | 73 | |
XM_011537620.1 | XP_011535922.1 | 10 - 11 | 1335 | 73 | |
XM_017009287.1 | XP_016864776.1 | 9 - 10 | 1322 | 73 | |
XM_017009288.1 | XP_016864777.1 | 11 - 12 | 1485 | 73 | |
XM_017009289.1 | XP_016864778.1 | 11 - 12 | 1478 | 73 | |
XM_017009290.1 | XP_016864779.1 | 10 - 11 | 1545 | 73 | |
XM_017009291.1 | XP_016864780.1 | 10 - 11 | 1238 | 73 | |
XM_017009292.1 | XP_016864781.1 | 10 - 11 | 1182 | 73 | |
GenBank mRNA | AK293269.1 | - | 8 - 9 | 1079 | 73 |
AK293390.1 | - | 9 - 10 | 1163 | 73 | |
AK298661.1 | - | 5 - 6 | 717 | 73 | |
AK299971.1 | - | 7 - 8 | 875 | 73 | |
BC007680.2 | - | 10 - 11 | 1240 | 73 | |
BC014982.1 | - | 10 - 11 | 1385 | 73 | |
U18937.1 | - | 10 - 11 | 1347 | 73 |