The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278731.1 | NP_001265660.1 | 7 - 8 | 975 | 79 |
NM_001278732.1 | NP_001265661.1 | 6 - 7 | 928 | 79 | |
NM_012208.3 | NP_036340.1 | 8 - 9 | 1050 | 79 | |
XM_011537619.1 | XP_011535921.1 | 9 - 10 | 1063 | 79 | |
XM_011537620.1 | XP_011535922.1 | 8 - 9 | 964 | 79 | |
XM_017009287.1 | XP_016864776.1 | 7 - 8 | 951 | 79 | |
XM_017009288.1 | XP_016864777.1 | 9 - 10 | 1114 | 79 | |
XM_017009289.1 | XP_016864778.1 | 9 - 10 | 1107 | 79 | |
XM_017009290.1 | XP_016864779.1 | 8 - 9 | 1174 | 79 | |
XM_017009291.1 | XP_016864780.1 | 8 - 9 | 867 | 79 | |
XM_017009292.1 | XP_016864781.1 | 8 - 9 | 811 | 79 | |
GenBank mRNA | AK293269.1 | - | 6 - 7 | 708 | 79 |
AK293390.1 | - | 7 - 8 | 792 | 79 | |
AK298661.1 | - | 3 - 4 | 346 | 79 | |
AK299971.1 | - | 5 - 6 | 504 | 79 | |
AK312976.1 | - | 8 - 9 | 868 | 79 | |
BC007680.2 | - | 8 - 9 | 869 | 79 | |
BC014982.1 | - | 8 - 9 | 1014 | 79 | |
BQ228600.1 | - | 6 - 7 | 662 | 79 | |
U18937.1 | - | 8 - 9 | 976 | 79 |