The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005733.2 | NP_005724.1 | 13 - 14 | 2179 | 96 |
GenBank mRNA | AF070672.1 | - | 12 - 13 | 1710 | 96 |
AF153329.1 | - | 13 - 14 | 1717 | 96 | |
AK024804.1 | - | 2 - 3 | 236 | 96 | |
AK025790.1 | - | 13 - 14 | 1738 | 96 | |
AK296879.1 | - | 14 - 15 | 1693 | 96 | |
BC012999.2 | - | 13 - 14 | 1717 | 96 |