Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF20A
Entrez Gene ID
10112
Gene Name
kinesin family member 20A
Gene Aliases
MKLP2, RAB6KIFL
Chromosome Location
Chr.5: 138178728 - 138187715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005733.2 NP_005724.1 3 - 4 752 70
GenBank mRNA AF070672.1 - 2 - 3 283 70
AF153329.1 - 3 - 4 290 70
AK025790.1 - 3 - 4 311 70
BC012999.2 - 3 - 4 290 70

View Full Product Details