The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033578.2 | NP_001028750.1 | 12 - 13 | 1429 | 136 |
NM_013257.4 | NP_037389.4 | 12 - 13 | 1152 | 136 | |
NM_170709.2 | NP_733827.2 | 12 - 13 | 1152 | 136 | |
GenBank mRNA | AF085233.2 | - | 11 - 12 | 136 | |
AF169035.1 | - | 11 - 12 | 913 | 136 | |
AK223523.1 | - | 12 - 13 | 1121 | 136 | |
BC015326.1 | - | 11 - 12 | 946 | 136 | |
CR536581.1 | - | 9 - 10 | 691 | 136 | |
CR933673.1 | - | 12 - 13 | 1086 | 136 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204173.1 | NP_001191102.1 | 14 - 15 | 1523 | 136 |