The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033578.2 | NP_001028750.1 | 9 - 10 | 1143 | 109 |
NM_013257.4 | NP_037389.4 | 9 - 10 | 866 | 109 | |
NM_170709.2 | NP_733827.2 | 9 - 10 | 866 | 109 | |
GenBank mRNA | AF169035.1 | - | 8 - 9 | 627 | 109 |
AK074666.1 | - | 9 - 10 | 820 | 109 | |
AK223523.1 | - | 9 - 10 | 835 | 109 | |
AK291428.1 | - | 9 - 10 | 1143 | 109 | |
BC015326.1 | - | 8 - 9 | 660 | 109 | |
CR536581.1 | - | 6 - 7 | 405 | 109 | |
CR933673.1 | - | 9 - 10 | 800 | 109 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204173.1 | NP_001191102.1 | 11 - 12 | 1237 | 109 |