The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018390.3 | NP_060860.1 | 7 - 7 | 4259 | 113 |
XM_006724866.2 | XP_006724929.1 | 7 - 7 | 4011 | 113 | |
GenBank mRNA | AK091006.1 | - | 1 - 1 | 1440 | 114 |
AK097346.1 | - | 1 - 1 | 697 | 112 |