The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005257514.1 | XP_005257571.1 | 1 - 1 | 119 | 69 |
GenBank mRNA | AK131210.1 | - | 1 - 1 | 547 | 69 |
BC110848.1 | - | 1 - 1 | 63 | 69 |