The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032271.2 | NP_115647.2 | 13 - 14 | 1358 | 92 |
XM_005255627.4 | XP_005255684.1 | 13 - 14 | 1398 | 92 | |
XM_011522700.1 | XP_011521002.1 | 13 - 14 | 1400 | 92 | |
GenBank mRNA | AK090624.1 | - | 13 - 14 | 1372 | 92 |
AL136921.1 | - | 12 - 13 | 1254 | 92 | |
AY569455.1 | - | 12 - 13 | 1311 | 92 | |
BC024267.1 | - | 11 - 12 | 1144 | 92 |