The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032271.2 | NP_115647.2 | 14 - 15 | 1441 | 53 |
XM_005255627.4 | XP_005255684.1 | 14 - 15 | 1481 | 53 | |
XM_011522700.1 | XP_011521002.1 | 14 - 15 | 1483 | 53 | |
GenBank mRNA | AK090624.1 | - | 14 - 15 | 1455 | 53 |
AL136921.1 | - | 13 - 14 | 1337 | 53 | |
AY569455.1 | - | 13 - 14 | 1394 | 53 | |
BC024267.1 | - | 12 - 13 | 1227 | 53 |