The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021074.4 | NP_066552.2 | 5 - 6 | 587 | 98 |
XM_017025782.1 | XP_016881271.1 | 5 - 6 | 777 | 98 | |
GenBank mRNA | AF147367.1 | - | 1 - 2 | 62 | 98 |
AK291865.1 | - | 5 - 6 | 483 | 98 | |
AK312129.1 | - | 5 - 6 | 505 | 98 | |
AW051350.1 | - | 3 - 4 | 98 | ||
BC001632.1 | - | 5 - 6 | 497 | 98 | |
BC017487.1 | - | 5 - 6 | 495 | 98 | |
BC071689.1 | - | 5 - 6 | 477 | 98 | |
BG742948.1 | - | 5 - 6 | 587 | 98 | |
CR456928.1 | - | 5 - 6 | 473 | 98 | |
M22538.1 | - | 5 - 6 | 491 | 98 |