Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHL1
Entrez Gene ID
10752
Gene Name
cell adhesion molecule L1 like
Gene Aliases
CALL, L1CAM2
Chromosome Location
Chr.3: 196588 - 409417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253387.1 NP_001240316.1 5 - 6 1029 98
NM_001253388.1 NP_001240317.1 3 - 4 481 98
NM_006614.3 NP_006605.2 5 - 6 1029 98
XM_006712938.1 XP_006713001.1 5 - 6 640 98
XM_006712939.3 XP_006713002.1 4 - 5 957 98
XM_006712940.3 XP_006713003.1 4 - 5 871 98
XM_011533292.1 XP_011531594.1 5 - 6 1010 98
XM_011533294.1 XP_011531596.1 5 - 6 640 98
XM_011533295.1 XP_011531597.1 5 - 6 1010 98
XM_011533296.1 XP_011531598.1 5 - 6 640 98
XM_017005566.1 XP_016861055.1 6 - 7 5534 98
XM_017005567.1 XP_016861056.1 4 - 5 742 98
XM_017005568.1 XP_016861057.1 4 - 5 549 98
XM_017005569.1 XP_016861058.1 4 - 5 957 98
XM_017005570.1 XP_016861059.1 4 - 5 871 98
XM_017005571.1 XP_016861060.1 4 - 5 742 98
XM_017005572.1 XP_016861061.1 5 - 6 1037 98
XM_017005573.1 XP_016861062.1 4 - 5 957 98
GenBank mRNA AB209329.1 - 5 - 6 854 98
AF002246.1 - 5 - 6 658 98
AK126878.1 - 5 - 6 1008 98
BC104918.1 - 3 - 4 418 98
BC143496.1 - 3 - 4 415 98

View Full Product Details