Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHL1
Entrez Gene ID
10752
Gene Name
cell adhesion molecule L1 like
Gene Aliases
CALL, L1CAM2
Chromosome Location
Chr.3: 196588 - 409417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253387.1 NP_001240316.1 9 - 10 1632 66
NM_001253388.1 NP_001240317.1 8 - 9 1132 66
NM_006614.3 NP_006605.2 10 - 11 1680 66
XM_006712938.1 XP_006713001.1 10 - 11 1291 66
XM_006712939.3 XP_006713002.1 9 - 10 1608 66
XM_006712940.3 XP_006713003.1 9 - 10 1522 66
XM_011533292.1 XP_011531594.1 10 - 11 1661 66
XM_011533294.1 XP_011531596.1 9 - 10 1243 66
XM_011533295.1 XP_011531597.1 9 - 10 1613 66
XM_011533296.1 XP_011531598.1 10 - 11 1291 66
XM_017005566.1 XP_016861055.1 11 - 12 6185 66
XM_017005567.1 XP_016861056.1 9 - 10 1393 66
XM_017005568.1 XP_016861057.1 9 - 10 1200 66
XM_017005569.1 XP_016861058.1 8 - 9 1560 66
XM_017005570.1 XP_016861059.1 8 - 9 1474 66
XM_017005571.1 XP_016861060.1 8 - 9 1345 66
XM_017005572.1 XP_016861061.1 10 - 11 1688 66
XM_017005573.1 XP_016861062.1 9 - 10 1608 66
GenBank mRNA AB209329.1 - 9 - 10 1457 66
AF002246.1 - 10 - 11 1309 66
AK126878.1 - 9 - 10 1611 66
BC104918.1 - 7 - 8 1021 66
BC143496.1 - 7 - 8 1018 66

View Full Product Details