Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRGC
Entrez Gene ID
56269
Gene Name
immunity related GTPase cinema
Gene Aliases
CINEMA, IFGGE, IRGC1, Iigp5, R30953_1
Chromosome Location
Chr.19: 43716062 - 43720021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005259058.3 XP_005259115.1 2 - 2 108 100

View Full Product Details