The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162429.2 | NP_001155901.1 | 5 - 6 | 767 | 88 |
NM_001256192.1 | NP_001243121.1 | 5 - 6 | 767 | 88 | |
NM_013374.5 | NP_037506.2 | 5 - 6 | 767 | 88 | |
XM_011533252.1 | XP_011531554.1 | 5 - 6 | 990 | 88 | |
XM_011533253.1 | XP_011531555.1 | 5 - 6 | 529 | 88 | |
XM_017005488.1 | XP_016860977.1 | 5 - 6 | 529 | 88 | |
GenBank mRNA | AF151793.1 | - | 5 - 6 | 735 | 88 |
AF250394.1 | - | 5 - 6 | 699 | 88 | |
AF349951.1 | - | 5 - 6 | 740 | 88 | |
AK295042.1 | - | 1 - 2 | 99 | 88 | |
BC020066.1 | - | 5 - 6 | 717 | 88 | |
BC068454.1 | - | 5 - 6 | 748 | 88 | |
BM669193.1 | - | 1 - 2 | 88 | ||
BT007367.1 | - | 5 - 6 | 609 | 88 | |
GQ131806.1 | - | 5 - 6 | 693 | 88 |