Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNOT10
Entrez Gene ID
25904
Gene Name
CCR4-NOT transcription complex subunit 10
Gene Aliases
-
Chromosome Location
Chr.3: 32685145 - 32773875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256741.1 NP_001243670.1 10 - 11 1530 65
NM_001256742.1 NP_001243671.1 10 - 11 1479 65
NM_015442.2 NP_056257.1 10 - 11 1530 65
NR_046352.1 9 - 10 1368 65
XM_006713084.3 XP_006713147.1 10 - 11 1486 65
XM_006713085.3 XP_006713148.1 10 - 11 1486 65
XM_011533566.2 XP_011531868.1 10 - 11 1486 65
XM_011533567.2 XP_011531869.1 6 - 7 747 65
XM_017006109.1 XP_016861598.1 10 - 11 1530 65
XM_017006110.1 XP_016861599.1 10 - 11 1530 65
XM_017006111.1 XP_016861600.1 10 - 11 1530 65
XM_017006112.1 XP_016861601.1 10 - 11 1530 65
GenBank mRNA AK021695.1 - 8 - 9 964 65
AK022576.1 - 10 - 11 1332 65
AK022952.1 - 10 - 11 1469 65
AK023227.1 - 10 - 11 1487 65
AK027026.1 - 7 - 8 972 65
AK297085.1 - 9 - 10 1105 65
AK298693.1 - 9 - 10 1168 65
AK302196.1 - 10 - 11 1471 65
AK309670.1 - 9 - 10 1293 65
BC002928.2 - 10 - 11 1432 65
BC002931.2 - 10 - 11 1438 65

View Full Product Details