The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142929.1 | NP_001136401.1 | 6 - 7 | 1081 | 94 |
NM_001142933.1 | NP_001136405.1 | 7 - 8 | 1151 | 94 | |
NM_001142934.1 | NP_001136406.1 | 7 - 8 | 1151 | 94 | |
NM_020549.4 | NP_065574.3 | 6 - 7 | 1081 | 94 | |
NM_020984.3 | NP_066264.3 | 6 - 7 | 747 | 94 | |
NM_020985.3 | NP_066265.3 | 7 - 8 | 909 | 94 | |
NM_020986.3 | NP_066266.3 | 6 - 7 | 737 | 94 | |
GenBank mRNA | AF305907.1 | - | 6 - 7 | 1081 | 94 |
AF305908.2 | - | 7 - 8 | 1151 | 94 | |
AF305909.1 | - | 6 - 7 | 747 | 94 | |
BC130615.1 | - | 5 - 6 | 607 | 94 | |
BC130617.1 | - | 5 - 6 | 607 | 94 | |
S45018.1 | - | 5 - 6 | 598 | 94 | |
S56138.1 | - | 6 - 7 | 1081 | 94 |