Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OXSR1
Entrez Gene ID
9943
Gene Name
oxidative stress responsive 1
Gene Aliases
OSR1
Chromosome Location
Chr.3: 38165051 - 38255488 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005109.2 NP_005100.1 17 - 18 1852 95
XM_005265638.2 XP_005265695.1 18 - 19 1993 95
XM_011534331.2 XP_011532633.1 17 - 18 1904 95
XM_017007601.1 XP_016863090.1 16 - 17 1659 95
XM_017007602.1 XP_016863091.1 19 - 20 2179 95
GenBank mRNA AB017642.1 - 17 - 18 1852 95
AB029024.1 - 17 - 18 1605 95
AF087893.1 - 12 - 13 998 95
BC008726.2 - 17 - 18 1835 95

View Full Product Details