Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMCO5A
Entrez Gene ID
145942
Gene Name
transmembrane and coiled-coil domains 5A
Gene Aliases
TMCO5
Chromosome Location
Chr.15: 37934607 - 37997541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152453.3 NP_689666.2 9 - 10 766 91
XM_005254168.3 XP_005254225.1 9 - 10 747 91
XM_011521262.2 XP_011519564.1 9 - 10 747 91
XM_011521263.1 XP_011519565.1 9 - 10 747 91
GenBank mRNA AK093126.1 - 9 - 10 747 91
BC029221.2 - 8 - 9 679 91

View Full Product Details