The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042494.1 | NP_001035959.1 | 17 - 18 | 2508 | 119 |
NM_001042495.1 | NP_001035960.1 | 17 - 18 | 2183 | 119 | |
NM_001042496.1 | NP_001035961.1 | 17 - 18 | 2574 | 119 | |
NM_001042497.1 | NP_001035962.1 | 15 - 16 | 2277 | 119 | |
NM_005135.2 | NP_005126.1 | 16 - 17 | 2147 | 119 | |
NM_133647.1 | NP_598408.1 | 16 - 17 | 2322 | 119 | |
XM_006720793.3 | XP_006720856.1 | 15 - 16 | 3057 | 119 | |
GenBank mRNA | AF105366.1 | - | 16 - 17 | 2322 | 119 |
AF108831.1 | - | 16 - 17 | 2056 | 119 | |
AF116242.1 | - | 16 - 17 | 2158 | 119 | |
AF477977.1 | - | 17 - 18 | 2515 | 119 | |
AF531258.1 | - | 15 - 16 | 2277 | 119 | |
AF531259.1 | - | 17 - 18 | 2515 | 119 | |
AF531260.1 | - | 17 - 18 | 2153 | 119 | |
AK128133.1 | - | 15 - 16 | 1830 | 119 | |
AK292550.1 | - | 17 - 18 | 2261 | 119 | |
AK315283.1 | - | 16 - 17 | 2119 | 119 | |
AL117500.1 | - | 13 - 14 | 1750 | 119 | |
BC051744.1 | - | 17 - 18 | 2511 | 119 | |
BC070107.1 | - | 17 - 18 | 2178 | 119 | |
BC126241.1 | - | 16 - 17 | 2055 | 119 | |
BC126243.1 | - | 16 - 17 | 2055 | 119 | |
BX648195.1 | - | 16 - 17 | 3264 | 119 | |
DQ138323.1 | - | 17 - 18 | 2573 | 119 |